3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetic acid 97%

Lieferant: Apollo Scientific

PC302831-1G
APOSPC302831-1GEA 0 EUR
APOSPC302831-1G
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetic acid 97%
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetic acid
Formel: C₉H₅ClF₄O₂
Molecular Weight: 256,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23795331
CAS-Nummer: 1431329-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR