3-Chlor-4-(trifluormethyl)benzamid 97%

Lieferant: Apollo Scientific

PC302832-5G PC302832-1G
APOSPC302832-5GEA 0 EUR
APOSPC302832-5G APOSPC302832-1G
3-Chlor-4-(trifluormethyl)benzamid 97%
3-Chlor-4-(trifluormethyl)benzamid
Formel: C₈H₅ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11519326
CAS-Nummer: 1092460-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR