3'-Chlor-5'-(pentafluorthio)acetophenon 97%

Lieferant: Apollo Scientific

PC302833-1G PC302833-5G PC302833-10G
APOSPC302833-1GEA 0 EUR
APOSPC302833-1G APOSPC302833-5G APOSPC302833-10G
3'-Chlor-5'-(pentafluorthio)acetophenon 97%
Formel: C₈H₆ClF₅OS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652455
CAS-Nummer: 1180486-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR