3-Chloro-5-(pentafluorothio)cinnamic acid 98%

Lieferant: Apollo Scientific

PC302835-1G
APOSPC302835-1GEA 0 EUR
APOSPC302835-1G
3-Chloro-5-(pentafluorothio)cinnamic acid 98%
3-Chloro-5-(pentafluorothio)cinnamic acid
Formel: C₉H₆ClF₅O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD16652454
CAS-Nummer: 1240261-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR