3-Fluor-2-methoxybenzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC302841-1G
APOSPC302841-1GEA 256 EUR
APOSPC302841-1G
3-Fluor-2-methoxybenzylbromid 97%
3-Fluor-2-methoxybenzylbromid
Formel: C₈H₈BrFO
Lagertemperatur: Raumtemperatur
MDL: MFCD09025370
CAS-Nummer: 916420-50-3
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR