3-Fluoro-5-(trifluoromethoxy)phenylacetonitrile 98%
Lieferant: Apollo Scientific
PC302849-1G
APOSPC302849-1GEA
229
EUR
APOSPC302849-1G
3-Fluoro-5-(trifluoromethoxy)phenylacetonitrile 98%
3-Fluoro-5-(trifluoromethoxy)phenylacetonitrile
Formel:
C₉H₅F₄NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD18783227 CAS-Nummer: 1352999-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...