4-Chlor-2-fluor-3-methoxybenzylamin 95%

Lieferant: Apollo Scientific
Gefahr

PC302861-1G
APOSPC302861-1GEA 145 EUR
APOSPC302861-1G
4-Chlor-2-fluor-3-methoxybenzylamin 95%
4-Chlor-2-fluor-3-methoxybenzylamin
Formel: C₈H₉ClFNO
Molecular Weight: 189,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687209
CAS-Nummer: 1323966-34-2
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR