4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine
PC302862-1G
APOSPC302862-1GEA
159
EUR
APOSPC302862-1G
4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine
4-Chloro-2-fluoro-3-methoxy-DL-phenylalanine
Formel:
C₁₀H₁₁ClFNO₃ Molecular Weight: 247,65 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19687211 CAS-Nummer: 1706418-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...