4-Chlor-3-ethoxy-2-fluoranilin 97%

Lieferant: Apollo Scientific

PC302865-1G PC302865-5G
APOSPC302865-1GEA 159 EUR
APOSPC302865-1G APOSPC302865-5G
4-Chlor-3-ethoxy-2-fluoranilin 97%
4-Chlor-3-ethoxy-2-fluoranilin
Formel: C₈H₉ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD19687218
CAS-Nummer: 1323966-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR