4-Chloro-3-ethoxy-2-fluorobenzonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-3-ethoxy-2-fluorobenzonitrile

PC302866-1G
APOSPC302866-1GEA 159 EUR
APOSPC302866-1G
4-Chloro-3-ethoxy-2-fluorobenzonitrile 97%
4-Chloro-3-ethoxy-2-fluorobenzonitrile
Formel: C₉H₇ClFNO
Molecular Weight: 199,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201019
CAS-Nummer: 1373920-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR