4-Chloro-3-ethoxy-2-fluorophenylacetonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-3-ethoxy-2-fluorophenylacetonitrile

PC302868-1G
APOSPC302868-1GEA 159 EUR
APOSPC302868-1G
4-Chloro-3-ethoxy-2-fluorophenylacetonitrile 98%
4-Chloro-3-ethoxy-2-fluorophenylacetonitrile
Formel: C₁₀H₉ClFNO
Molecular Weight: 213,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687187
CAS-Nummer: 1323966-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR