4-Ethoxy-3,5-difluorobenzamide 97%

Lieferant: Apollo Scientific

PC302874-1G
APOSPC302874-1GEA 50.9 EUR
APOSPC302874-1G
4-Ethoxy-3,5-difluorobenzamide 97%
4-Ethoxy-3,5-difluorobenzamide
Formel: C₉H₉F₂NO₂
Molecular Weight: 201,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258685
CAS-Nummer: 1017779-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR