4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
Lieferant: Apollo Scientific
Synonyme:
4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
PC302877-1G
APOSPC302877-1GEA
229
EUR
APOSPC302877-1G
4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
4-Fluoro-2-trifluoromethoxy-DL-phenylalanine
Formel:
C₁₀H₉F₄NO₃ Molecular Weight: 267,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19687192 CAS-Nummer: 1435806-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...