4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride 97%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride

PC302881-1G
APOSPC302881-1GEA 87 EUR
APOSPC302881-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride
Formel: C₉H₄ClF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD22201065
CAS-Nummer: 1373920-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR