4-Methoxy-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302883-10G PC302883-5G PC302883-1G
APOSPC302883-10GEA 282 EUR
APOSPC302883-10G APOSPC302883-5G APOSPC302883-1G
4-Methoxy-2-(trifluormethyl)benzylbromid 98%
4-Methoxy-2-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09025406
CAS-Nummer: 916420-86-5
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR