4-Methoxy-3-trifluoromethoxy-DL-phenylalanine

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-3-trifluoromethoxy-DL-phenylalanine

PC302884-1G
APOSPC302884-1GEA 235 EUR
APOSPC302884-1G
4-Methoxy-3-trifluoromethoxy-DL-phenylalanine
4-Methoxy-3-trifluoromethoxy-DL-phenylalanine
Formel: C₁₁H₁₂F₃NO₄
Molecular Weight: 279,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09915503
CAS-Nummer: 1259979-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR