4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile

Lieferant: Apollo Scientific

PC302885-1G
APOSPC302885-1GEA 316 EUR
APOSPC302885-1G
4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile
4-Methoxy-3-(trifluoromethoxy)phenylacetonitrile
Formel: C₁₀H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18398529
CAS-Nummer: 853771-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR