4-Methoxy-3-methyl-5-(trifluoromethyl)benzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzaldehyde, 4-methoxy-3-methyl-5-(trifluoromethyl)-

PC302886-5G PC302886-1G
APOSPC302886-5GEA 224 EUR
APOSPC302886-5G APOSPC302886-1G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzaldehyde 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzaldehyde
Formel: C₁₀H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22682818
CAS-Nummer: 1431329-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR