4-Methyl-3-(trifluormethyl)benzaldehyd

Lieferant: Apollo Scientific
Achtung

PC302888-5G PC302888-10G PC302888-1G
APOSPC302888-5GEA 557 EUR
APOSPC302888-5G APOSPC302888-10G APOSPC302888-1G
4-Methyl-3-(trifluormethyl)benzaldehyd
4-Methyl-3-(trifluoromethyl)benzaldehyde
Formel: C₉H₇F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04116033
CAS-Nummer: 93249-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR