6-Chloro-2,3-difluorobenzamide

Lieferant: Apollo Scientific

Synonyme: 6-chloro-2,3-difluorobenzamide

PC302897-5G
APOSPC302897-5GEA 348 EUR
APOSPC302897-5G
6-Chloro-2,3-difluorobenzamide
6-Chloro-2,3-difluorobenzamide
Formel: C₇H₄ClF₂NO
Molecular Weight: 191.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201031
CAS-Nummer: 1373920-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR