6-Ethoxy-2,3-difluorobenzoic acid 97%

Lieferant: Apollo Scientific

PC302901-1G
APOSPC302901-1GEA 115 EUR
APOSPC302901-1G
6-Ethoxy-2,3-difluorobenzoic acid 97%
6-Ethoxy-2,3-difluorobenzoic acid
Formel: C₉H₈F₂O₃
Molecular Weight: 202,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23795338
CAS-Nummer: 1431329-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR