2-Chlor-4,6-bis(trifluormethyl)benzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC302910-1G PC302910-5G PC302910-10G
APOSPC302910-1GEA 0 EUR
APOSPC302910-1G APOSPC302910-5G APOSPC302910-10G
2-Chlor-4,6-bis(trifluormethyl)benzaldehyd 98%
2-Chlor-4,6-bis(trifluormethyl)benzaldehyd
Formel: C₉H₃ClF₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD09025366
CAS-Nummer: 916420-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR