2-Chlor-4,6-bis(trifluormethyl)benzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302910-1G
PC302910-5G
PC302910-10G
APOSPC302910-1GEA
0
EUR
APOSPC302910-1G
APOSPC302910-5G
APOSPC302910-10G
2-Chlor-4,6-bis(trifluormethyl)benzaldehyd 98%
2-Chlor-4,6-bis(trifluormethyl)benzaldehyd
Formel:
C₉H₃ClF₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025366 CAS-Nummer: 916420-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...