2-Chlor-4,6-bis(trifluormethyl)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302911-1G
PC302911-5G
PC302911-10G
APOSPC302911-1GEA
0
EUR
APOSPC302911-1G
APOSPC302911-5G
APOSPC302911-10G
2-Chlor-4,6-bis(trifluormethyl)benzoesäure 98%
2-Chlor-4,6-bis(trifluormethyl)benzoesäure
Formel:
C₉H₃ClF₆O₂ Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025365 CAS-Nummer: 916420-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...