2-Fluoro-4-(pentafluorothio)phenylacetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-4-(pentafluorosulphur)phenylacetic acid

PC302913-1G
APOSPC302913-1GEA 348 EUR
APOSPC302913-1G
2-Fluoro-4-(pentafluorothio)phenylacetic acid 97%
2-Fluoro-4-(pentafluorothio)phenylacetic acid
Formel: C₈H₆F₆O₂S
Molecular Weight: 280,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652509
CAS-Nummer: 1240257-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR