2-Fluoro-4,6-bis(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

PC302917-1G
APOSPC302917-1GEA 58 EUR
APOSPC302917-1G
2-Fluoro-4,6-bis(trifluoromethyl)benzamide 97%
2-Fluoro-4,6-bis(trifluoromethyl)benzamide
Formel: C₉H₄F₇NO
Molecular Weight: 275,123 g/mol
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258671
CAS-Nummer: 1017778-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR