2-Fluor-5-methyl-4-(trifluormethyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

PC302921-1G PC302921-5G
APOSPC302921-1GEA 183 EUR
APOSPC302921-1G APOSPC302921-5G
2-Fluor-5-methyl-4-(trifluormethyl)benzoesäure
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19687178
CAS-Nummer: 1323955-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR