2-Methoxy-3-(trifluoromethyl)benzoyl chloride 97%
Lieferant: Apollo Scientific
PC302925-1G
APOSPC302925-1GEA
79.9
EUR
APOSPC302925-1G
2-Methoxy-3-(trifluoromethyl)benzoyl chloride 97%
2-Methoxy-3-(trifluoromethyl)benzoyl chloride
Formel:
C₉H₆ClF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832312 CAS-Nummer: 1017778-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...