2-Methoxy-5-(trifluormethoxy)phenol 98%

Lieferant: Apollo Scientific
Achtung

PC302927-10G PC302927-5G PC302927-25G
APOSPC302927-10GEA 0 EUR
APOSPC302927-10G APOSPC302927-5G APOSPC302927-25G
2-Methoxy-5-(trifluormethoxy)phenol 98%
2-Methoxy-5-(trifluormethoxy)phenol
Formel: C₈H₇F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09832306
CAS-Nummer: 895572-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR