2-Methoxy-5-(trifluoromethyl)phenol 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-5-trifluoromethylphenol

PC302930-250MG
APOSPC302930-250MGEA 72.5 EUR
APOSPC302930-250MG
2-Methoxy-5-(trifluoromethyl)phenol 98%
2-Methoxy-5-(trifluoromethyl)phenol
Formel: C₈H₇F₃O₂
Molecular Weight: 192,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832299
CAS-Nummer: 349-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR