2-Methoxy-6-(trifluormethyl)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC302931-10G
PC302931-5G
PC302931-25G
PC302931-100G
APOSPC302931-10GEA
0
EUR
APOSPC302931-10G
APOSPC302931-5G
APOSPC302931-25G
APOSPC302931-100G
2-Methoxy-6-(trifluormethyl)benzaldehyd
2-Methoxy-6-(trifluoromethyl)benzaldehyde
Formel:
C₉H₇F₃O₂ Schmelzpunkt: 46…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832324 CAS-Nummer: 1017778-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...