2'-Methyl-3'-(trifluormethyl)acetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Methyl-3-(trifluoromethyl)phenyl)ethanone

PC302932-10G PC302932-5G PC302932-1G
APOSPC302932-10GEA 328 EUR
APOSPC302932-10G APOSPC302932-5G APOSPC302932-1G
2'-Methyl-3'-(trifluormethyl)acetophenon 98%
2'-Methyl-3'-(trifluormethyl)acetophenon
Formel: C₁₀H₉F₃O
Molecular Weight: 202.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832283
CAS-Nummer: 1017778-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR