2'-Methyl-3'-(trifluormethyl)acetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Methyl-3-(trifluoromethyl)phenyl)ethanone
PC302932-10G
PC302932-5G
PC302932-1G
APOSPC302932-10GEA
328
EUR
APOSPC302932-10G
APOSPC302932-5G
APOSPC302932-1G
2'-Methyl-3'-(trifluormethyl)acetophenon 98%
2'-Methyl-3'-(trifluormethyl)acetophenon
Formel:
C₁₀H₉F₃O Molecular Weight: 202.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832283 CAS-Nummer: 1017778-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...