3-(2,3,4-Trifluorophenyl)propionic acid 97%

Lieferant: Apollo Scientific

PC302935-1G
APOSPC302935-1GEA 87 EUR
APOSPC302935-1G
3-(2,3,4-Trifluorophenyl)propionic acid 97%
3-(2,3,4-Trifluorophenyl)propionic acid
Formel: C₉H₇F₃O₂
Molecular Weight: 204,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660214
CAS-Nummer: 377084-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR