3,5-Difluoro-2-methoxybenzamide 97%

Lieferant: Apollo Scientific

PC302939-5G
APOSPC302939-5GEA 180 EUR
APOSPC302939-5G
3,5-Difluoro-2-methoxybenzamide 97%
3,5-Difluoro-2-methoxybenzamide
Formel: C₈H₇F₂NO₂
Molecular Weight: 187,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06409086
CAS-Nummer: 874804-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR