3-Chloro-4-ethoxy-5-fluorocinnamic acid 97%

Lieferant: Apollo Scientific

PC302946-1G
APOSPC302946-1GEA 168 EUR
APOSPC302946-1G
3-Chloro-4-ethoxy-5-fluorocinnamic acid 97%
3-Chloro-4-ethoxy-5-fluorocinnamic acid
Formel: C₁₁H₁₀ClFO₃
Molecular Weight: 244,647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832348
CAS-Nummer: 1017779-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR