3-Chloro-5-(trifluoromethoxy)benzoyl chloride

Lieferant: Apollo Scientific

PC302947-10G PC302947-5G
APOSPC302947-10GEA 0 EUR
APOSPC302947-10G APOSPC302947-5G
3-Chloro-5-(trifluoromethoxy)benzoyl chloride
3-Chloro-5-(trifluoromethoxy)benzoyl chloride
Formel: C₈H₃Cl₂F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660307
CAS-Nummer: 886503-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR