3-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302960-10G PC302960-5G PC302960-1G
APOSPC302960-10GEA 0 EUR
APOSPC302960-10G APOSPC302960-5G APOSPC302960-1G
3-Methoxy-4-(trifluoromethyl)benzoyl chloride 97%
3-Methoxy-4-(trifluoromethyl)benzoyl chloride
Formel: C₉H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18398587
CAS-Nummer: 1261571-92-9
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR