3-Methyl-5-(trifluormethoxy)benzamid 97%

Lieferant: Apollo Scientific

PC302963-10G PC302963-5G PC302963-1G
APOSPC302963-10GEA 449 EUR
APOSPC302963-10G APOSPC302963-5G APOSPC302963-1G
3-Methyl-5-(trifluormethoxy)benzamid 97%
3-Methyl-5-(trifluormethoxy)benzamid
Formel: C₉H₈F₃NO₂
Molecular Weight: 219.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025372
CAS-Nummer: 916420-53-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR