3-Methyl-5-(trifluoromethoxy)phenylacetonitrile 98%
Lieferant: Apollo Scientific
PC302965-1G
APOSPC302965-1GEA
237
EUR
APOSPC302965-1G
3-Methyl-5-(trifluoromethoxy)phenylacetonitrile 98%
3-Methyl-5-(trifluoromethoxy)phenylacetonitrile
Formel:
C₁₀H₈F₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025374 CAS-Nummer: 916420-56-9 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...