4'-(Pentafluorthio)acetophenon 98%

Lieferant: Apollo Scientific
Achtung

PC302967-5G PC302967-1G PC302967-25G
APOSPC302967-5GEA 0 EUR
APOSPC302967-5G APOSPC302967-1G APOSPC302967-25G
4'-(Pentafluorthio)acetophenon 98%
4'-(Pentafluorthio)acetophenon
Formel: C₈H₇F₅OS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652408
CAS-Nummer: 401892-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR