4-Chloro-3-(trifluoromethoxy)benzyl alcohol 97%

Lieferant: Apollo Scientific

PC302978-10G PC302978-5G PC302978-25G
APOSPC302978-10GEA 190 EUR
APOSPC302978-10G APOSPC302978-5G APOSPC302978-25G
4-Chloro-3-(trifluoromethoxy)benzyl alcohol 97%
4-Chloro-3-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660248
CAS-Nummer: 886500-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR