4-Chloro-3-ethoxy-2-fluorophenylacetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-3-ethoxy-2-fluorophenylacetic acid

PC302986-1G
APOSPC302986-1GEA 180 EUR
APOSPC302986-1G
4-Chloro-3-ethoxy-2-fluorophenylacetic acid 98%
4-Chloro-3-ethoxy-2-fluorophenylacetic acid
Formel: C₁₀H₁₀ClFO₃
Molecular Weight: 232,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687188
CAS-Nummer: 1323955-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR