4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%

Lieferant: Apollo Scientific
Achtung

PC302990-1G
APOSPC302990-1GEA 271 EUR
APOSPC302990-1G
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid 98%
4-Fluoro-2-(trifluoromethoxy)phenylacetic acid
Formel: C₉H₆F₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19687177
CAS-Nummer: 1323966-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR