4-Fluor-3-(trifluormethoxy)phenylessigsäure 97%

Lieferant: Apollo Scientific
Achtung

PC302992-1G PC302992-5G
APOSPC302992-1GEA 0 EUR
APOSPC302992-1G APOSPC302992-5G
4-Fluor-3-(trifluormethoxy)phenylessigsäure 97%
4-Fluor-3-(trifluormethoxy)phenylessigsäure
Formel: C₉H₆F₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06660258
CAS-Nummer: 886501-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR