4-Methoxy-3-(trifluoromethoxy)phenylacetic acid

Lieferant: Apollo Scientific

PC302998-1G
APOSPC302998-1GEA 0 EUR
APOSPC302998-1G
4-Methoxy-3-(trifluoromethoxy)phenylacetic acid
4-Methoxy-3-(trifluoromethoxy)phenylacetic acid
Formel: C₁₀H₉F₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD18398521
CAS-Nummer: 1261557-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR