5-Fluor-2-(trifluormethoxy)benzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
PC303003-10G
PC303003-5G
PC303003-1G
APOSPC303003-10GEA
337
EUR
APOSPC303003-10G
APOSPC303003-5G
APOSPC303003-1G
5-Fluor-2-(trifluormethoxy)benzoesäure
5-Fluor-2-(trifluormethoxy)benzoesäure
Formel:
C₈H₄F₄O₃ Molecular Weight: 224.11 g/mol Schmelzpunkt: 89…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519333 CAS-Nummer: 1092460-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...