6-Chloro-2,3-difluorobenzyl alcohol 97%

Lieferant: Apollo Scientific

Synonyme: (6-CHLORO-2,3-DIFLUOROPHENYL)METHANOL, 6-Chloro-2,3-difluorobenzenemethanol

PC303009-1G
APOSPC303009-1GEA 98.6 EUR
APOSPC303009-1G
6-Chloro-2,3-difluorobenzyl alcohol 97%
6-Chloro-2,3-difluorobenzyl alcohol
Formel: C₇H₅ClF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07368108
CAS-Nummer: 887585-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR