2,6-Dichloro-4-(pentafluorosulphur)nitrobenzene 97%

Lieferant: Apollo Scientific

PC303021-1G
APOSPC303021-1GEA 1250 EUR
APOSPC303021-1G
2,6-Dichloro-4-(pentafluorosulphur)nitrobenzene 97%
2,6-Dichloro-4-(pentafluorosulphur)nitrobenzene
Formel: C₆H₂Cl₂F₅NO₂S
Molecular Weight: 318.049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23795321
CAS-Nummer: 1431329-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR