2,6-Difluor-4-methoxybenzylamin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2,6-Difluoro-4-methoxyphenyl)methanamine

PC303024-1G
APOSPC303024-1GEA 130 EUR
APOSPC303024-1G
2,6-Difluor-4-methoxybenzylamin 98%
2,6-Difluor-4-methoxybenzylamin
Formel: C₈H₉F₂NO
Molecular Weight: 173,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115924
CAS-Nummer: 771573-20-7
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR